missing translation for 'onlineSavingsMsg'
Learn More

9,9'-Spirobifluorene, 98%

Product Code. 15441199
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441199 1 g 1g
15461199 250 mg 250mg
15451199 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441199 Supplier Thermo Scientific Alfa Aesar Supplier No. H64133.03
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 159-66-0 | C25H16 | 316.403 g/mol

A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A blue light emitting material in organic light emitting diodes (OLEDs). Also used as a blue-emitting material in electroluminescent devices.

Solubility
Soluble in common organic solvents. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159-66-0
Molecular Formula C25H16
Molecular Weight (g/mol) 316.403
MDL Number MFCD08704216
InChI Key SNFCXVRWFNAHQX-UHFFFAOYSA-N
Synonym 9,9'-spirobifluorene, 9,9'-spirobi fluorene, 9,9'-spirobi 9h-fluorene, 9,9-spirobi 9h-fluorene, 9,9-spirobifluorene, pubchem3102, 9.9'-spirobifluorene, acmc-209dk2, 9,9 inverted exclamation mark-spirobi-fluorene
PubChem CID 135975
IUPAC Name 9,9'-spirobi[fluorene]
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=CC=CC=C5C6=CC=CC=C46

Specifications

Melting Point 195°C to 200°C
Color White to Yellow
Quantity 1 g
Beilstein 1914463
Solubility Information Soluble in common organic solvents. Slightly soluble in water.
Formula Weight 316.4
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 9,9'-Spirobifluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.