missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,9'-Spirobifluorene-2-boronic acid, 98%, Thermo Scientific Chemicals
POA - £252.00
Chemical Identifiers
CAS | 236389-21-2 |
---|---|
Molecular Formula | C25H17BO2 |
Molecular Weight (g/mol) | 360.22 |
MDL Number | MFCD13248568 |
InChI Key | WDDLHUWVLROJLA-UHFFFAOYSA-N |
Synonym | 9,9'-spirobi fluoren-2-ylboronic acid, 9,9'-spirobi 9h-fluorene-2-boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, 9,9'-spirobi fluorene-2-ylboronic acid, boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl, 9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, boronic acid, 9,9'-spirobi 9h-fluoren-2-yl, acmc-209g4k, 9,9'-spirobifluorene-2-boronic acid, 9,9'-spirobi fluorene-2-yl boronic acid |
PubChem CID | 22564369 |
IUPAC Name | 9,9'-spirobi[fluorene]-2-ylboronic acid |
SMILES | OB(O)C1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15461779
|
Thermo Scientific Alfa Aesar
H64525.MD |
250 mg | |||||||
15441779
|
Thermo Scientific Alfa Aesar
H64525.03 |
1 g |
£64.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15451779
|
Thermo Scientific Alfa Aesar
H64525.06 |
5 g |
£252.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
236389-21-2 | |
360.22 | |
WDDLHUWVLROJLA-UHFFFAOYSA-N | |
22564369 | |
OB(O)C1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12 |
C25H17BO2 | |
MFCD13248568 | |
9,9'-spirobi fluoren-2-ylboronic acid, 9,9'-spirobi 9h-fluorene-2-boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, 9,9'-spirobi fluorene-2-ylboronic acid, boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl, 9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, boronic acid, 9,9'-spirobi 9h-fluoren-2-yl, acmc-209g4k, 9,9'-spirobifluorene-2-boronic acid, 9,9'-spirobi fluorene-2-yl boronic acid | |
9,9'-spirobi[fluorene]-2-ylboronic acid |
Specifications
236389-21-2 | |
C25H17BO2 | |
10478712 | |
WDDLHUWVLROJLA-UHFFFAOYSA-N | |
9,9'-spirobi[fluorene]-2-ylboronic acid | |
22564369 | |
98% | |
9,9'-Spirobifluorene-2-boronic acid |
White | |
MFCD13248568 | |
9,9'-spirobi fluoren-2-ylboronic acid, 9,9'-spirobi 9h-fluorene-2-boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, 9,9'-spirobi fluorene-2-ylboronic acid, boronic acid, b-9,9'-spirobi 9h-fluoren-2'-yl, 9,9'-spirobi 9h-fluoren-2'-yl-boronic acid, boronic acid, 9,9'-spirobi 9h-fluoren-2-yl, acmc-209g4k, 9,9'-spirobifluorene-2-boronic acid, 9,9'-spirobi fluorene-2-yl boronic acid | |
OB(O)C1=CC2=C(C=C1)C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12 | |
360.22 | |
360.22 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only