missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethylxanthene, 98+%, Thermo Scientific Chemicals

Product Code. 11372439
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372439 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11372439 Supplier Thermo Scientific Alfa Aesar Supplier No. H27340.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19814-75-6 | Molecular Formula: C15H14O | Molecular Weight: 210.28 g/mol

9,9-Dimethylxanthene is suitable for use in the preparation of 9,9-dimethylxanthene-4,5-dicarboxylic acid. It may be used in the preparation of 9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene. It may be used in the preparation of 4,6-bis(dimesitylboryl)dibenzofuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,9-Dimethylxanthene is suitable for use in the preparation of 9,9-dimethylxanthene-4,5-dicarboxylic acid. It may be used in the preparation of 9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene. It may be used in the preparation of 4,6-bis(dimesitylboryl)dibenzofuran.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19814-75-6
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00134434
InChI Key MTVNAPYHLASOSX-UHFFFAOYSA-N
Synonym 9,9-dimethyl-9h-xanthene, 9h-xanthene, 9,9-dimethyl, 9,9-dimethylxantene, 9,9-dimethyl xanthene, xanthene, 9,9-dimethyl, ksc182m0n, 9,9-dimethylxanthene, 9,9-dimethyl-9h-xanthene #
PubChem CID 606997
SMILES CC1(C)C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 40°C to 43°C
Boiling Point 114°C to 115°C (0.6 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Solubility Information Insoluble in water.
IUPAC Name 9,9-dimethyl-9H-xanthene
Formula Weight 210.28
Percent Purity ≥98%
Chemical Name or Material 9,9-Dimethylxanthene

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.