missing translation for 'onlineSavingsMsg'
Learn More

9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester, 95%

Product Code. 15469465
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15469465 5 g 5g
15459465 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469465 Supplier Thermo Scientific Alfa Aesar Supplier No. H51954.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 325129-69-9 | C27H36B2O4 | 446.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 325129-69-9
Molecular Formula C27H36B2O4
Molecular Weight (g/mol) 446.201
MDL Number MFCD08704230
InChI Key RVFLMSKITNJVRB-UHFFFAOYSA-N
Synonym 2,2'-9,9-dimethyl-9h-fluorene-2,7-diyl bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2,7-diboronic acid bis pinacol ester, 2-9,9-dimethyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb256, 2,2-9,9-dimethyl-9h-fluorene-2,7-diyl, 9,9-dimethyl-9h-fluoren-2,7-diyl bisboronic acid pinacol ester, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9,9-dimethyl-9h-fluorene
PubChem CID 20640927
IUPAC Name 2-[9,9-dimethyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C3(C)C)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Quantity 5 g
Formula Weight 446.2
Percent Purity 95%
Chemical Name or Material 9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.