missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Hydroxybutyl)guanine, TRC

Product Code. 30395026
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395026 500 mg
30390866 50 mg
30397255 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395026 Supplier Toronto Research Chemicals Supplier No. TRCH231690500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23169-37-1
Molecular Formula C9H13N5O2
Molecular Weight (g/mol) 223.23
Synonym 2-Amino-1,9-dihydro-9-(4-hydroxybutyl)-6H-purin-6-one, 2-Amino-9-(4-hydroxybutyl)-6,9-dihydro-1H-purin-6-one, 2-Amino-9-(4-hydroxybutyl)-9H-purin-6-ol
IUPAC Name 2-amino-9-(4-hydroxybutyl)-1H-purin-6-one
SMILES NC1=Nc2c(ncn2CCCCO)C(=O)N1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H13N5O2/c10-9-12-7-6(8(16)13-9)11-5-14(7)3-1-2-4-15/h5,15H,1-4H2,(H3,10,12,13,16)
Formula Weight 223.1069
Chemical Name or Material 9-(4-Hydroxybutyl)guanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.