missing translation for 'onlineSavingsMsg'
Learn More

9,21-Dichloro-11β-hydroxy-16β-methyl-3,20-dioxopregna-1,4-dien-17-yl Furan-2-carboxylate (16-epi-Mometasone Furoate), Mikromol™

Product Code. 30051172
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051172 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051172 Supplier Mikromol Supplier No. MM1272.180025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

9,21-Dichloro-11β-hydroxy-16β-methyl-3,20-dioxopregna-1,4-dien-17-yl Furan-2-carboxylate (16-epi-Mometasone Furoate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 16-epi Mometasone Furoate
CAS 2231764-75-1
Analyte or Component Names 16-epi Mometasone Furoate
Shipping Condition Room Temperature
Molecular Formula C27H30Cl2O6
Certifications/Compliance ISO 9001
Synonym 9,21-Dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione, Mometasone Impurity S, Pregna-1,4-diene-3,20-dione, 9,21-dichloro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-, (11β,16β)-, 9,21-Dichloro-11β-hydroxy-16β-methyl-3,20-dioxopregna-1,4-dien-17-yl Furan-2-carboxylate, 16-epi-Mometasone Furoate, Mometasone Furoate Imp. S (EP)
InChI Formula InChI=1S/C27H30Cl2O6/c1-15-11-19-18-7-6-16-12-17(30)8-9-24(16,2)26(18,29)21(31)13-25(19,3)27(15,22(32)14-28)35-23(33)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,31H,6-7,11,13-14H2,1-3H3/t15-,18-,19-,21-,24-,25-,26-,27-/m0/s1
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)CCl
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-9-chloro-17-(2-chloroacetyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
Molecular Weight (g/mol) 521.43
Quantity 25 mg
Formula Weight 520.1419 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.