missing translation for 'onlineSavingsMsg'
Learn More

9-(2,3-Dideoxy-beta-D-glycero-pentofuranosyl)-9H-purin-6-amine (2',3'-Dideoxyadenosine), Mikromol™

Product Code. 30050291
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30050291

Brand: Mikromol MM1047.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

9-(2,3-Dideoxy-beta-D-glycero-pentofuranosyl)-9H-purin-6-amine (2',3'-Dideoxyadenosine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2',3'-Dideoxyadenosine
CAS 4097-22-7
Analyte or Component Names 2',3'-Dideoxyadenosine
Shipping Condition Room Temperature
Molecular Formula C10 H13 N5 O2
Certifications/Compliance ISO 9001
Synonym Didanosine Imp. G (EP), Didanosine USP Related Compound B, Didanosine USP RC B, Adenosine, 2',3'-dideoxy-, 2',3'-Dideoxyadenosine, Dideoxyadenosine, NSC 98700, 9-(2,3-Dideoxy-beta-D-glycero-pentofuranosyl)-9H-purin-6-amine, Didanosine Related Compound B, Didanosine Impurity G
InChI Formula InChI=1S/C10H13N5O2/c11-9-8-10(13-4-12-9)15(5-14-8)7-2-1-6(3-16)17-7/h4-7,16H,1-3H2,(H2,11,12,13)/t6-,7+/m0/s1
SMILES Nc1ncnc2c1ncn2[C@H]3CC[C@@H](CO)O3
IUPAC Name [(2S,5R)-5-(6-aminopurin-9-yl)oxolan-2-yl]methanol
Molecular Weight (g/mol) 235.243
Quantity 100 mg
Formula Weight 235.107 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.