missing translation for 'onlineSavingsMsg'
Learn More

9,11α-Epoxy-17-hydroxy-7α-(methoxycarbonyl)-3-oxo-17α-pregn-4-ene-21-carboxylic Acid Potassium Salt, Mikromol

Product Code. 30461049
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461049 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461049 Supplier Mikromol Supplier No. MM3530.070025
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

9,11α-Epoxy-17-hydroxy-7α-(methoxycarbonyl)-3-oxo-17α-pregn-4-ene-21-carboxylic Acid Potassium Salt, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Eplerenone Hydroxyacid Potassium Salt
CAS 95716-98-6
Analyte or Component Names Eplerenone Hydroxyacid Potassium Salt
Shipping Condition Room Temperature
Molecular Formula C24 H31 O7 . K
Certifications/Compliance ISO 9001
InChI Formula InChI=1S/C24H32O7.K/c1-21-7-4-14(25)10-13(21)11-15(20(28)30-3)19-16-5-8-23(29,9-6-18(26)27)22(16,2)12-17-24(19,21)31-17;/h10,15-17,19,29H,4-9,11-12H2,1-3H3,(H,26,27);/q;+1/p-1/t15-,16+,17-,19+,21+,22+,23-,24-;/m1./s1
Solution Type Neat
SMILES [K+].COC(=O)[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@@]34O[C@@H]3C[C@@]5(C)[C@@H](CC[C@@]5(O)CCC(=O)[O-])[C@H]14
Molecular Weight (g/mol) 470.6
Quantity 25 mg
Formula Weight 470.1707 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.