missing translation for 'onlineSavingsMsg'
Learn More

9(10H)-Acridone, 99%

Product Code. 11472248
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472248 25 g 25g
11304629 1 g 1g
11462248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472248 Supplier Thermo Scientific Alfa Aesar Supplier No. A15222.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 578-95-0 | C13H9NO | 195.221 g/mol

9(10H)-Acridone is used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate, use for dyeing the organic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9(10H)-Acridone is used in the preparation of methyl 9,10-dihydro-9-oxoacridine-10-pentanoate, use for dyeing the organic materials.

Solubility
Insoluble in benzene, chloroform, ether, water and ethanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578-95-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00005019
InChI Key FZEYVTFCMJSGMP-UHFFFAOYSA-N
Synonym acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine
PubChem CID 2015
ChEBI CHEBI:50756
IUPAC Name 10H-acridin-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2

Specifications

Melting Point 360°C to 362°C
Quantity 25 g
Beilstein 7104
Solubility Information Insoluble in benzene,chloroform,ether,water and ethanol.
Formula Weight 195.22
Percent Purity 99%
Chemical Name or Material 9(10H)-Acridone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.