missing translation for 'onlineSavingsMsg'
Learn More

9,10-Phenanthrenequinone, 95%, Thermo Scientific Chemicals

Catalog Number 11344629
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
84-11-7
C14H8O2
208.216
MFCD00001163
YYVYAPXYZVYDHN-UHFFFAOYSA-N
9,10-phenanthrenequinone, phenanthrenequinone, 9,10-phenanthrenedione, 9,10-phenanthraquinone, phenanthraquinone, 9,10-phenanthroquinone, 9-10 phenanthrene quinone, unii-42l7bz8h74, ccris 7615, phenanthrene, 9,10-dihydro-9,10-dioxo
6763
CHEBI:37454
phenanthrene-9,10-dione
C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)C2=O
This item is not returnable. View return policy
84-11-7
C14H8O2
208.216
MFCD00001163
YYVYAPXYZVYDHN-UHFFFAOYSA-N
9,10-phenanthrenequinone, phenanthrenequinone, 9,10-phenanthrenedione, 9,10-phenanthraquinone, phenanthraquinone, 9,10-phenanthroquinone, 9-10 phenanthrene quinone, unii-42l7bz8h74, ccris 7615, phenanthrene, 9,10-dihydro-9,10-dioxo
6763
CHEBI:37454
phenanthrene-9,10-dione
C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)C2=O

9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Phenanthrenequinon may be used for high quality passivation on silicon (100) surfaces. Quinones may serve as substrates for a variety of flavoenzymes, useful fluorescent compound and quinone. Also used in dyes, as a hardener for dental restoration, for the determination of copper by spectrophotometric method, and for organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 207°C to 212°C
Density 1.405
Boiling Point >360°C
Flash Point 245°C (473°F)
Quantity 5 g
UN Number UN3077
Beilstein 608838
Merck Index 14,7213
Solubility Information Insoluble in water.
Formula Weight 208.22
Percent Purity 95%
Chemical Name or Material 9,10-Phenanthrenequinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'