missing translation for 'onlineSavingsMsg'
Learn More

9,10-Ethanoanthracene-9(10H)-propanamide, TRC

Product Code. 30368821
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368821 50 mg 50mg
30380002 25 mg 25mg
30369821 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368821 Supplier Toronto Research Chemicals Supplier No. TRCE67655550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9,10-Ethanoanthracene-9(10H)-propanamide
Analyte or Component Names 9,10-Ethanoanthracene-9(10H)-propanamide
Shipping Condition Room Temperature
Molecular Formula C19 H19 N O
Synonym 9,10-Ethanoanthracene-9(10H)-propionamide
InChI Formula InChI=1S/C19H19NO/c20-18(21)10-12-19-11-9-13(14-5-1-3-7-16(14)19)15-6-2-4-8-17(15)19/h1-8,13H,9-12H2,(H2,20,21)
Solution Type Neat
SMILES NC(=O)CCC12CCC(c3ccccc13)c4ccccc24
IUPAC Name 3-(1-tetracyclo[6.6.2.02,7.09,14]hexadeca-2(7),3,5,9,11,13-hexaenyl)propanamide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 277.36
Quantity 50 mg
Formula Weight 277.147
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.