missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydroxystearic Acid, Pract., MP Biomedicals

Product Code. 11477734 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477734 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477734 Supplier MP Biomedicals Supplier No. 0520304005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 120-87-6
Molecular Formula C18H36O4
MDL Number MFCD00046726
Quantity 5 mg
Synonym 9,10-dihydroxystearic acid, dioxystearinsaeure, 9,10-dihydroxystearate, 9,10-dihydroxystearinsaeure, 9,10-dihydroxy-octadecanoic acid, rac threo-9,10-dihydroxystearic acid, dhsa, parent, 9,10-dihydroxy stearic acid, 9,10-dhsa
InChI Key VACHUYIREGFMSP-IAGOWNOFSA-N
SMILES CCCCCCCC[C@@H](O)[C@H](O)CCCCCCCC(O)=O
IUPAC Name (9R,10R)-9,10-dihydroxyoctadecanoic acid
Molecular Weight (g/mol) 316.48
PubChem CID 89377
ChEBI CHEBI:28724
Formula Weight 316.26
Chemical Name or Material 9, 10-Dihydroxystearic Acid, Pract.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.