missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydroergocristine Methanesulfonate, TRC

Product Code. 30381103
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381103 5 mg -
30387604 25 mg -
30386926 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381103 Supplier Toronto Research Chemicals Supplier No. TRCD4464405MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24730-10-7
Molecular Formula C35 H41 N5 O5 . C H4 O3 S
Molecular Weight (g/mol) 707.84
Synonym Dihydroergocristine Mesilate, (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-(1-methylethyl)-3,6-dioxo-octahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate, (5'α,10α)-9,10-Dihydro-12'-hydroxy-2'-(1-methylethyl)-5'-(phenylmethyl)ergotaman-3',6',18-trione methanesulfonate, Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-(1-methylethyl)-5'-(phenylmethyl)-, (5'α,10α)-, methanesulfonate (1:1) (ACI), Ergocristine, 9,10-dihydro-, monomethanesulfonate (salt) (8CI), Ergocristine, dihydro-, methanesulfonate (6CI, 7CI), Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-(1-methylethyl)-5'-(phenylmethyl)-, (5'α,10α)-, monomethanesulfonate (salt) (9CI), 9,10-Dihydroergocristine methanesulfonate, Dihydroergocristine mesylate, Dihydroergocristine methanesulfonate, GNF Pf 3462
IUPAC Name (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-(1-methylethyl)-3,6-dioxo-octahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate
SMILES CC(C)[C@@]1(NC(=O)[C@@H]2C[C@H]3[C@@H](Cc4c[nH]c5cccc3c45)N(C)C2)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C1=O.CS(=O)(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C35H41N5O5.CH4O3S/c1-20(2)34(37-31(41)23-16-25-24-11-7-12-26-30(24)22(18-36-26)17-27(25)38(3)19-23)33(43)40-28(15-21-9-5-4-6-10-21)32(42)39-14-8-13-29(39)35(40,44)45-34;1-5(2,3)4/h4-7,9-12,18,20,23,25,27-29,36,44H,8,13-17,19H2,1-3H3,(H,37,41);1H3,(H,2,3,4)/t23-,25-,27-,28+,29+,34-,35+;/m1./s1
Formula Weight 707.2989
Chemical Name or Material Dihydroergocristine Mesilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.