missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydro-1-benzo[a]pyrene-7(8H)-one Oxime, TRC

Product Code. 30383704
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383704 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383704 Supplier Toronto Research Chemicals Supplier No. TRCD44842510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9,10-Dihydro-1-benzo[a]pyrene-7(8H)-one Oxime
CAS 88909-82-4
Molecular Formula C20 H15 N O
Quantity 10 mg
Synonym Benzo[a]pyren-7(8H)-one, 9,10-dihydro-, oxime, 9,10-Dihydro-1-benzo[a]pyrene-7(8H)-one oxime
InChI Formula InChI=1S/C20H15NO/c22-21-18-6-2-5-15-16-10-9-13-4-1-3-12-7-8-14(11-17(15)18)20(16)19(12)13/h1,3-4,7-11,22H,2,5-6H2/b21-18+
SMILES O\N=C\1/CCCc2c1cc3ccc4cccc5ccc2c3c45
IUPAC Name (NE)-N-(9,10-dihydro-8H-benzo[a]pyren-7-ylidene)hydroxylamine
Molecular Weight (g/mol) 285.34
Formula Weight 285.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.