missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dichloroanthracene, TRC

Product Code. 30388603
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
Product Code. Quantity unitSize
30388603 100 mg -
30369873 50 mg 50mg
30383426 10 mg -
3 options
This item is not returnable. View return policy

Product Code. 30388603

Brand: Toronto Research Chemicals TRCD435943100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-48-1
Formula Weight 246.0003
InChI Formula InChI=1S/C14H8Cl2/c15-13-9-5-1-2-6-10(9)14(16)12-8-4-3-7-11(12)13/h1-8H
IUPAC Name 9,10-dichloroanthracene
Molecular Formula C14H8Cl2
Molecular Weight (g/mol) 247.12
SMILES Clc1c2ccccc2c(Cl)c3ccccc13
Synonym 9,10-Dichloroanthracene, Anthracene, 9,10-dichloro-, NSC 42963, 9,10-Dichloroanthracene, Anthracene, 9,10-dichloro-, NSC 42963
Chemical Name or Material 9,10-Dichloroanthracene

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.