missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dibromoanthracene-2-sulfonic Acid Sodium Salt, TRC

Product Code. 30379130
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379130 500 mg
30378891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379130 Supplier Toronto Research Chemicals Supplier No. TRCD423000500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87796-18-7
Molecular Formula C14 H7 Br2 O3 S . Na
Molecular Weight (g/mol) 438.07
IUPAC Name sodium;9,10-dibromoanthracene-2-sulfonate
SMILES [Na+].[O-]S(=O)(=O)c1ccc2c(Br)c3ccccc3c(Br)c2c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H8Br2O3S.Na/c15-13-9-3-1-2-4-10(9)14(16)12-7-8(20(17,18)19)5-6-11(12)13;/h1-7H,(H,17,18,19);/q;+1/p-1
Formula Weight 435.838
Chemical Name or Material 9,10-Dibromoanthracene-2-sulfonic Acid, Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.