Learn More
9,10-Bis(phenylethynyl)anthracene
Description
9,10-Bis(phenylethynyl)anthracene is used as a chemiluminescent fluorophore with high quantum efficiency. It is used in lightsticks as a fluorophor producing ghostly green light. It is also used as a dopant for organic semiconductors in OLEDs. It is used as a reagent for chemiluminescence research. 9,10-Bis(phenylethynyl)anthracene is known to exert fluorescence properties due to its polycyclic aromatic hydrocarbon system and it is commonly used as an scintillator additive. 9,10-Bis(phenylethynyl)anthracene was also studied extensively as a fluorescence emitters for peroxyoxalate chemiluminescence.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 254°C to 255°C |
| Quantity | 5 g |
| Beilstein | 1891432 |
| Sensitivity | Light sensitive |
| Solubility Information | It is insoluble in water but is soluble in most organic solvents such as carbon disulfide,alcohols,benzene,chloroform,and hydronaphthalenes. |
| Formula Weight | 378.47 |
| Physical Form | Crystalline |
| Chemical Name or Material | 9,10-Bis (phenylethynyl)anthracene |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.