missing translation for 'onlineSavingsMsg'
Learn More

9,10-Anthraquinone, 98+%, Thermo Scientific Chemicals

Product Code. 11400693
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400693 250 g 250g
11490683 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400693 Supplier Thermo Scientific Alfa Aesar Supplier No. A12735.30
 Restricted Item
In Stock
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 84-65-1 | Molecular Formula: C14H8O2 | Molecular Weight: 208.22 g/mol

Anthraquinone is used in paper industry as a catalyst to increase the pulp production yield and to improves the fiber strength through reduction reaction of cellulose to carboxylic acid. It is also used as a precursor for dye formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthraquinone is used in paper industry as a catalyst to increase the pulp production yield and to improves the fiber strength through reduction reaction of cellulose to carboxylic acid. It is also used as a precursor for dye formation.

Solubility
Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 84-65-1
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.22
MDL Number MFCD00001188
InChI Key RZVHIXYEVGDQDX-UHFFFAOYSA-N
Synonym anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone
PubChem CID 6780
ChEBI CHEBI:40448
IUPAC Name anthracene-9,10-dione
SMILES O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12

Spécification

Melting Point 283°C to 287°C
Density 1.438
Boiling Point 377°C
Flash Point 185°C (365°F)
Odor Odorless
Quantity 250 g
Beilstein 390030
Merck Index 14,687
Solubility Information Insoluble in water.
Formula Weight 208.22
Percent Purity ≥98%
Chemical Name or Material 9,10-Anthraquinone
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.