missing translation for 'onlineSavingsMsg'
Learn More

9,10-12,13-Diepoxyoctadecanoic Acid, TRC

Product Code. 30380563
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380563 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380563 Supplier Toronto Research Chemicals Supplier No. TRCD44185510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9,10-12,13-Diepoxyoctadecanoic Acid
CAS 3012-69-9
Molecular Formula C18 H32 O4
Quantity 10 mg
Synonym 9,10:12,13-Diepoxyoctadecanoic Acid, 3-[(3-Pentyl-2-oxiranyl)methyl]-2-oxiraneoctanoic acid, 9,10:12,13-Diepoxyoctadecanoic acid, 9,12-Diepoxyoctadecanoic acid, Diepoxylinoleic acid, Linoleic acid dioxide, NSC 3581
InChI Formula InChI=1S/C18H32O4/c1-2-3-7-10-14-16(21-14)13-17-15(22-17)11-8-5-4-6-9-12-18(19)20/h14-17H,2-13H2,1H3,(H,19,20)
SMILES CCCCCC1OC1CC2OC2CCCCCCCC(=O)O
IUPAC Name 8-[3-[(3-pentyloxiran-2-yl)methyl]oxiran-2-yl]octanoic acid
Molecular Weight (g/mol) 312.444
Formula Weight 312.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.