missing translation for 'onlineSavingsMsg'
Learn More

(8R,9S)-Talazoparib, MedChemExpress

Product Code. 16469527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469527 Supplier Medchem Express Supplier No. HY16106A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (8R,9S)-Talazoparib ((8R,9S)-BMN-673) is an enantiomer of Talazoparib. (8R,9S)-Talazoparib is an PARP1 inhibitor, with an IC50 of 144 nM.

(8R,9S)-Talazoparib ((8R,9S)-BMN-673) is an enantiomer of Talazoparib. (8R,9S)-Talazoparib is an PARP1 inhibitor, with an IC50 of 144 nM.

Specifications

Chemical Name or Material (8R,9S)-Talazoparib
CAS 1207456-00-5
Color White
Purity Grade Notes Research
Molecular Formula C19H14F2N6O
Quantity 5 mg
Synonym (8R,9S)-BMN-673 (8R,9S)-LT-673
Solubility Information DMSO : ≥ 50 mg/mL (131.46 mM)
SMILES O=C1NN=C2C3=C1C=C(F)C=C3N[C@@H](C4=CC=C(F)C=C4)[C@@H]2C5=NC=NN5C
Molecular Weight (g/mol) 380.35
Formula Weight 380.35
Percent Purity 95.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.