missing translation for 'onlineSavingsMsg'
Learn More

8Beta-Hydroxy-10-oxo-8,14-dihydrothebaine, TRC

Product Code. 30391409
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391409 2.5 mg 2.5mg
30390654 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391409 Supplier Toronto Research Chemicals Supplier No. TRCH9508352.5MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155051-97-1
Molecular Formula C19H21NO5
Molecular Weight (g/mol) 343.37
Synonym (5alpha,8beta)-6,7-didehydro-4,5-epoxy-8-hydroxy-3,6-dimethoxy-17-methyl-morphinan-10-one
IUPAC Name (4S,5R,7aR,12bS)-5-hydroxy-7,9-dimethoxy-3-methyl-1,2,4,4a,5,7a-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-13-one
SMILES COC1=C[C@H](O)C2[C@@H]3N(C)CC[C@]24[C@H]1Oc5c(OC)ccc(C3=O)c45

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C19H21NO5/c1-20-7-6-19-13-9-4-5-11(23-2)17(13)25-18(19)12(24-3)8-10(21)14(19)15(20)16(9)22/h4-5,8,10,14-15,18,21H,6-7H2,1-3H3/t10-,14?,15-,18-,19+/m0/s1
Formula Weight 343.142
Chemical Name or Material 8beta-Hydroxy-10-oxo-8,14-dihydrothebaine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.