missing translation for 'onlineSavingsMsg'
Learn More

Tuttnauer 85L Vertical Autoclave

Product Code. 17861087 Shop All Tuttnauer Products
Click to view available options
Features:
Single Phase, 208 V
Single Phase, 230 V
Three Phase
Three Phase, Cooling
Three Phase, Cooling, Vacuum, Steam Generator, BioHazard Filter (CL3 Waste), Printer
Three Phase, Cooling, Vacuum, Steam Generator, Printer
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17861087

Brand: Tuttnauer 3870ELV208D

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The 3870ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.

Features

  • Fo software control
  • PID (Proportional Integral Differential) pressure control
  • Stores the last 200 cycles in built-in memory
  • Two independent flexible PT100 temperature sensors to prevent over-boiling of liquids and explosions of bottles
  • Up to 6 temperature sensors and 4 pressure sensors can be connected
  • 30 Identification Codes and Passwords for access level control
  • The controller and software comply with the 21 CFR part 11 standard

Innovative Multi-Color Display

  • Multi-color display for easier reading
  • Color is used to indicate the stage of the cycle
  • Quick access to important information
  • 26 Languages
  • Built-in view of historical cycle data

Specifications

Capacity (English) 18.7 gal.
Capacity (Metric) 85 L
Depth (Metric) Chamber 70 cm
Diameter (English) Chamber 15.2 in.
Depth (English) Chamber 28 in.
Diameter (Metric) Chamber 38 cm
Length (English) 31.6 in.
Length (Metric) 54 cm
Product Type Autoclave, Top-loading
Temperature (English) 221°F to 280°F
Temperature (Metric) 105°C to 138°C
Features Single Phase, 208 V
Material 316L Stainless Steel
Voltage 208 V
Sterility Non-sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.