Learn More
Moldex™ 8000 Particulate Filters
Description
Moldex 8000 Series P2 R D filters (8070). P2 R D particulate filters for the 8000 Series half mask. For protection against harmful and carcinogenic dusts, fumes and aerosols based on water and oil. 10 x WEL.
Moldex 8000 Series P3 R D filters (8080). P3 R D particulate filters for the 8000 Series half mask. For protection against harmful and carcinogenic dusts, fumes and aerosols including radioactive particulates as well as airborne biological substances and enzymes. 20 x WEL.
Moldex 8000 Series P3 R D filters (8080). P3 R D particulate filters for the 8000 Series half mask. For protection against harmful and carcinogenic dusts, fumes and aerosols including radioactive particulates as well as airborne biological substances and enzymes. 20 x WEL.
Specifications
Specifications
| Filter Type | P2RD |
| Product Type | Particulate Filter |
| For Use With (Equipment) | For Series 8000 Respiratory Filter |
| Quantity | 8/Pk. |