missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ 8-Well Tip Combs Magnetic Rod Systems

Product Code. 18081091
Click to view available options
Quantity:
50 Pieces
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 18081091

Brand: Macherey-Nagel™ 744960

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

8-well tip combs for magnetic rod systems.

  • The 8-well magnetic tip combs lock into position inside the instruments processing chamber
  • The tip combs are ideal for magnetic bead processing; the special design enables excellent recovery of magnetic beads

Specifications

Product Type Tip Comb
Content And Storage 50 Pieces
For Use With (Application) Tip combs for magnetic rod systems
For Use With (Equipment) Magnetic Beads
Quantity 50 Pieces
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.