missing translation for 'onlineSavingsMsg'
Learn More

8-(Trifluoromethyl)quinoline, 98%, Thermo Scientific Chemicals

Catalog Number p-7053670
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
317-57-7
C10H6F3N
197.16
MFCD09744668
AJXUSUNIYLSPER-UHFFFAOYSA-N
12647568
8-(trifluoromethyl)quinoline
C1=CC2=C(C(=C1)C(F)(F)F)N=CC=C2
This item is not returnable. View return policy
317-57-7
C10H6F3N
197.16
MFCD09744668
AJXUSUNIYLSPER-UHFFFAOYSA-N
12647568
8-(trifluoromethyl)quinoline
C1=CC2=C(C(=C1)C(F)(F)F)N=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Synonym 8-trifluoromethyl quinoline, 8-cf3 quinoline, acmc-1ag4x, quinoline,8-trifluoromethyl, quinoline, 8-trifluoromethyl
Formula Weight 197.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 8-(Trifluoromethyl)quinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'