missing translation for 'onlineSavingsMsg'
Learn More

8-Quinolinesulfonyl chloride, 98%

Product Code. 15555866 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15555866

Brand: Thermo Scientific Acros 132210050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18704-37-5 | C9H6ClNO2S | 227.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18704-37-5
Molecular Formula C9H6ClNO2S
Molecular Weight (g/mol) 227.67
InChI Key JUYUYCIJACTHMK-UHFFFAOYSA-N
Synonym 8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp
PubChem CID 29220
IUPAC Name quinoline-8-sulfonyl chloride
SMILES C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2

Specifications

Melting Point 126.0°C to 130.0°C
Boiling Point 306.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Quantity 5 g
Beilstein 22,V,7,567
Solubility Information Solubility in water: reacts.
Formula Weight 227.67
Percent Purity 98%
Chemical Name or Material 8-Quinolinesulfonyl chloride
compliance-icons
Product Identifier
  • Quinoline-8-sulphonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.