missing translation for 'onlineSavingsMsg'
Learn More

8-Quinolinecarboxylic acid, 98%, Thermo Scientific™

Product Code. 10191142 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10191142 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10191142 Supplier Thermo Scientific Acros Supplier No. 368802500
 Restricted Item
Estimated Shipment: 10-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 86-59-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 183°C to 185°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H7NO2
MDL Number MFCD00047619
Quantity 250 mg
Beilstein 22, 81
Fieser 06,450
Merck Index 15, 8187
Synonym 8-quinolinecarboxylic acid, 8-carboxyquinoline, 8-quinolinecarboxylicacid, unii-5xw75tfd94, ccris 6956, pubchem5868, acmc-209qal, 8-quinlinecarboxylic acid, 8-quinoline carboxylic acid, 8-quinoline-carboxylic acid
Solubility Information Solubility in water: insoluble
InChI Key QRDZFPUVLYEQTA-UHFFFAOYSA-N
SMILES C1=CC2=C(C(=C1)C(=O)O)N=CC=C2
IUPAC Name quinoline-8-carboxylic acid
Molecular Weight (g/mol) 173.17
PubChem CID 66582
Formula Weight 173.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 8-Quinolinecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 8-Quinolinecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.