Learn More
8-Methylquinoline, 97%, Thermo Scientific Chemicals
£37.10 - £143.00
Chemical Identifiers
CAS | 611-32-5 |
---|---|
Molecular Formula | C10H9N |
Molecular Weight (g/mol) | 143.19 |
MDL Number | MFCD00006810 |
InChI Key | JRLTTZUODKEYDH-UHFFFAOYSA-N |
Synonym | quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888 |
PubChem CID | 11910 |
ChEBI | CHEBI:48984 |
IUPAC Name | 8-methylquinoline |
SMILES | CC1=C2N=CC=CC2=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10037832
|
Thermo Scientific Acros
303620050 |
5 g | Glass bottle |
£37.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10752491
|
Thermo Scientific Acros
303620250 |
25 g | Glass bottle |
£143.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
611-32-5 | |
143.19 | |
JRLTTZUODKEYDH-UHFFFAOYSA-N | |
11910 | |
8-methylquinoline |
C10H9N | |
MFCD00006810 | |
quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888 | |
CHEBI:48984 | |
CC1=C2N=CC=CC2=CC=C1 |
Specifications
611-32-5 | |
100.0 | |
Yellow | |
245°C to 248°C | |
Authentic | |
C10H9N | |
MFCD00006810 | |
quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888 | |
CC1=C2N=CC=CC2=CC=C1 | |
143.19 | |
CHEBI:48984 | |
97% | |
8-Methylquinoline, 97% |
96.0 | |
-80°C | |
1.0500g/mL | |
104°C | |
96% min. (GC) | |
1.6140 to 1.6160 (20°C, 589nm) | |
1.05 | |
JRLTTZUODKEYDH-UHFFFAOYSA-N | |
8-methylquinoline | |
11910 | |
143.19 | |
Liquid |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 210-264-0
missing translation for 'rtecsNumber' : VC0562000
missing translation for 'tsca' : TSCA
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only