missing translation for 'onlineSavingsMsg'
Learn More

8-Methyl Varenicline Dihydrochloride, TRC

Product Code. 30455108
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455108 1 g 1g
30450414 500 mg 500mg
30454547 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455108 Supplier Toronto Research Chemicals Supplier No. TRCT3101701G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Methyl Varenicline Dihydrochloride
Molecular Formula C14H15N3 • 2HCl
Quantity 1 g
Synonym 8-Methyl-7,8,9,10-tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline Dihydrochloride
InChI Formula InChI=1S/C14H15N3.2ClH/c1-17-7-9-4-10(8-17)12-6-14-13(5-11(9)12)15-2-3-16-14;;/h2-3,5-6,9-10H,4,7-8H2,1H3;2*1H
SMILES CN1CC2CC(C3=CC4=NC=CN=C4C=C23)C1.Cl.Cl
IUPAC Name 8-methyl-7,8,9,10-tetrahydro-6H-6,10-methanoazepino[4,5-g]quinoxaline dihydrochloride
Molecular Weight (g/mol) 298.21
Formula Weight 297.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.