missing translation for 'onlineSavingsMsg'
Learn More

8-Methyl-8-azabicyclo[3.2.1]octan-3-one oxime hydrochloride, 97%, Thermo Scientific™

Product Code. 10425302
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10425302 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10425302 Supplier Thermo Scientific Maybridge Supplier No. BTB10716DA
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 212079-30-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H15ClN2O
MDL Number MFCD01073570
Quantity 1 g
Synonym 8-methyl-8-azabicyclo 3.2.1 octan-3-one oxime hydrochloride, n-8-methyl-8-azabicyclo 3.2.1 octan-3-ylidene hydroxylamine hydrochloride, 8-methyl-8-azabicyclo3.2.1octan-3-one oxime hydrochloride, n-8-methyl-8-azabicyclo 3.2.1 octan-3-ylidene hydroxylamine-hydrogen chloride 1/1, n-3e-8-methyl-8-azabicyclo 3.2.1 octan-3-ylidene hydroxylamine hydrochloride
InChI Key XEGNNKBCZMXBOA-UHFFFAOYNA-N
SMILES Cl.CN1C2CCC1CC(C2)=NO
IUPAC Name N-(8-methyl-8-azabicyclo[3.2.1]octan-3-ylidene)hydroxylamine;hydrochloride
Molecular Weight (g/mol) 190.67
PubChem CID 2737441
Formula Weight 190.67
Percent Purity 97%
Chemical Name or Material 8-Methyl-8-azabicyclo[3.2.1]octan-3-one oxime hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.