missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxypsoralen, 99%

Product Code. 10368490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10368490 1 g Glass bottle 1g
10328540 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10368490 Supplier Thermo Scientific Acros Supplier No. 214150010
  • ONLINE EXCLUSIVE PRICE
    £35.40
    £29.65 / 1g
    Save £5.75
    16% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 298-81-7 | C12H8O4 | 216.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 298-81-7
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
MDL Number MFCD00005009
InChI Key QXKHYNVANLEOEG-UHFFFAOYSA-N
Synonym methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin
PubChem CID 4114
ChEBI CHEBI:18358
IUPAC Name 9-methoxyfuro[3,2-g]chromen-7-one
SMILES COC1=C2OC=CC2=CC2=C1OC(=O)C=C2

Spécification

CAS Min % 98.5
CAS Max % 100.0
Melting Point 143.0°C to 148.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (on anhydrous substance) (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 19, I, 711
Merck Index 15, 6059
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: sparingly soluble in boiling water,sparingly soluble in ether,soluble in boiling alcohol,acetone,acetic acid,,propylene glycol and benzene,freely soluble in chloroform
Formula Weight 216.19
Percent Purity 99%
Physical Form Needle-like Crystals or Powder
Chemical Name or Material 8-Methoxypsoralen, 99%
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • 8-Methoxypsoralen
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 4
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1A
  • Skin corrosion/irritation Category 1 B
Mention de danger
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.