missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxypsoralen, 98%

Product Code. 11488976
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488976 1 g 1g
11498976 5 g 5g
11408986 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488976 Supplier Thermo Scientific Alfa Aesar Supplier No. A17108.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 298-81-7 | C12H8O4 | 216.19 g/mol

8-Methoxypsoralen, is used in Photochemotherapy (methoxsalen with long wave ultraviolet radiation) is indicated for the repigmentation of idiopathic vitiligo. It is also used in Photopheresis (methoxsalen with long wave ultraviolet radiation of white blood cells) is indicated for use with the UVAR* System in the palliative treatment of the skin manifestations of cutaneous T-cell lymphoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Methoxypsoralen, is used in Photochemotherapy (methoxsalen with long wave ultraviolet radiation) is indicated for the repigmentation of idiopathic vitiligo. It is also used in Photopheresis (methoxsalen with long wave ultraviolet radiation of white blood cells) is indicated for use with the UVAR* System in the palliative treatment of the skin manifestations of cutaneous T-cell lymphoma.

Solubility
Soluble in acetone, ethanol (slight), acetic acid, and chloroform (50 mg/ml). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-81-7
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
MDL Number MFCD00005009
InChI Key QXKHYNVANLEOEG-UHFFFAOYSA-N
Synonym methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin
PubChem CID 4114
ChEBI CHEBI:18358
IUPAC Name 9-methoxyfuro[3,2-g]chromen-7-one
SMILES COC1=C2OC=CC2=CC2=C1OC(=O)C=C2

Specifications

Melting Point 146°C to 150°C
Quantity 1 g
Beilstein 196453
Sensitivity Light sensitive
Merck Index 14,5988
Solubility Information Soluble in acetone,ethanol (slight),acetic acid,and chloroform (50mg/ml). Insoluble in water.
Formula Weight 216.2
Percent Purity 99%
Chemical Name or Material 8-Methoxypsoralen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.