missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxy Psoralen, TRC

Product Code. 30473193
Change view
Click to view available options
Quantity:
25 mg
250 mg
5 g
Unit Size:
250mg
25mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473193 25 mg 25mg
30481534 5 g 5g
30475182 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473193 Supplier Toronto Research Chemicals Supplier No. TRCM26079525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-81-7
Molecular Formula C12 H8 O4
Molecular Weight (g/mol) 216.19
Synonym 5-Benzofuranacrylic acid, 6-hydroxy-7-methoxy-, δ-lactone (7CI), 9-Methoxy-7H-furo[3,2-g][1]benzopyran-7-one, 5-Demethoxyisoimpinellin, 8-MOP, 8-MP, 8-Methoxy-6,7-furanocoumarin, 8-Methoxy[furano-3',2':6,7-coumarin], 8-Methoxypsoralen, 8-Methoxypsoralene, Ammodin, Ammoidin, Deltapsoralen, Geroxalen, Meladinin, Meladinine, Meladoxen, Meloxine, Methoxa-Dome, Methoxsalen, Methoxsalene, NSC 45923, New-Meladinin, Oxsoralen, Oxsoralen Lotion, Oxsoralen-Ultra, Oxypsoralen, Puvalen, Puvamet, Uvadex, Vitpso, Xanthotoxin, Xanthotoxine
IUPAC Name 9-methoxyfuro[3,2-g]chromen-7-one
SMILES COc1c2OC(=O)C=Cc2cc3ccoc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H8O4/c1-14-12-10-8(4-5-15-10)6-7-2-3-9(13)16-11(7)12/h2-6H,1H3
Formula Weight 216.0423
Chemical Name or Material Xanthotoxin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.