missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 98%

Product Code. 11363217
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363217 1 g 1g
11373217 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363217 Supplier Thermo Scientific Alfa Aesar Supplier No. L13751.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32178-63-5 | C12H14O3 | 206.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32178-63-5
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD01075657
InChI Key VOAFGTBETRJDPU-UHFFFAOYSA-N
Synonym 8-methoxy-1,2,3,4-tetrahydronaphtalene-2-carboxylic acid, 1,2,3,4-tetrahydro-8-methoxynaphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid, 8-methoxy-1,2,3,4-tetrahydrophtalene-2-carboxylic acid, 2-naphthalenecarboxylicacid, 1,2,3,4-tetrahydro-8-methoxy
PubChem CID 3624709
IUPAC Name 8-methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid
SMILES COC1=CC=CC2=C1CC(CC2)C(=O)O

Specifications

Melting Point 141°C to 143°C
Quantity 1 g
Beilstein 2269890
Formula Weight 206.24
Percent Purity 98%
Chemical Name or Material 8-Methoxy-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.