missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline (White to Light-Buff Needles or Powder), Fisher BioReagents

Product Code. 10113243
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10113243 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10113243 Supplier Fisher BioReagents Supplier No. BP436100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
8-Hydroxyquinoline is used to stabilize phenol.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Hydroxyquinoline
CAS 148-24-3
Melting Point 72°C
Color White Beige Yellow
Boiling Point 267°C
Ignition Residue 0.05% max.
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Molecular Formula C9H7NO
MDL Number MFCD00006807
Quantity 100 g
Merck Index 15, 4881
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
Solubility Information 0.05% max. Insoluble in Alcohol
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
SMILES OC1=C2N=CC=CC2=CC=C1
IUPAC Name quinolin-8-ol
Molecular Weight (g/mol) 145.16
PubChem CID 1923
ChEBI CHEBI:48981
Formula Weight 145.16
Vapor Pressure 4.7hPa at 100°C
Percent Purity ≥99.0%
Physical Form Solid
Chemical Name or Material 8-Hydroxyquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 8-Hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.