missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline sulfate monohydrate, 98%

Product Code. 10665002 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10665002 100 g Glass bottle 100g
10533121 500 g Glass bottle 500g
10237113 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10665002 Supplier Thermo Scientific Acros Supplier No. 148451000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207386-91-2 | 0·5 H2O | 203.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207386-91-2
Molecular Formula 0·5 H2O
Molecular Weight (g/mol) 203.2
MDL Number MFCD00149610
InChI Key BNCXJZDIJIVJJO-UHFFFAOYSA-N
Synonym 8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 182.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Merck Index 14, 4843
Solubility Information Solubility in water: soluble in water. Other solubilities: slightly soluble in alcohol, insoluble in ether
Formula Weight 203.2
Percent Purity 98%
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 8-Hydroxyquinoline sulfate monohydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 8-Hydroxyquinoline sulfate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.