missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline sulfate monohydrate, 98%

Product Code. 11357815
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357815 25 g 25g
11367815 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357815 Supplier Thermo Scientific Alfa Aesar Supplier No. L02743.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207386-91-2 | C18H18N2O7S | 406.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207386-91-2
Molecular Formula C18H18N2O7S
Molecular Weight (g/mol) 406.41
MDL Number MFCD00149610
InChI Key BNCXJZDIJIVJJO-UHFFFAOYSA-N
Synonym 8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard

Specifications

Melting Point 176°C to 179°C
Quantity 25 g
Beilstein 3760550
Formula Weight 406.42 (388.40 Anhydrous)
Percent Purity 99%
Chemical Name or Material 8-Hydroxyquinoline sulfate monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.