missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, MedChemExpress

Product Code. 16458556
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458556 Supplier Medchem Express Supplier No. HYB1005
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 8-Hydroxyquinoline (8-Hydroxyquinoline) is a monoprotic bidentate chelating agent, exhibits antiseptic, disinfectant, and pesticide properties, functioning as a transcription inhibitor.

8-Hydroxyquinoline (8-Hydroxyquinoline) is a monoprotic bidentate chelating agent, exhibits antiseptic, disinfectant, and pesticide properties, functioning as a transcription inhibitor.

Specifications

Chemical Name or Material 8-Hydroxyquinoline
CAS 148-24-3
Color Tan
Purity Grade Notes Research
Molecular Formula C9H7NO
Quantity 100 mg
Synonym 8-Quinolinol
Solubility Information DMSO : 50 mg/mL (344.45 mM; Need ultrasonic)
SMILES OC1=C2N=CC=CC2=CC=C1
Molecular Weight (g/mol) 145.16
Formula Weight 145.16
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.