missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, ACS reagent, Thermo Scientific Chemicals

Product Code. 10132780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
1kg
2.5kg
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10132780 500 g Glass bottle 500g
10639653 25 g Glass bottle 25g
10274730 100 g Glass bottle 100g
10297933 1 kg Glass bottle 1kg
10669833 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10132780 Supplier Thermo Scientific Acros Supplier No. 121905000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148-24-3
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006807
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
PubChem CID 1923
ChEBI CHEBI:48981
IUPAC Name quinolin-8-ol
SMILES OC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point 72.0°C to 74.0°C
Color Beige or White to Yellow
Boiling Point 267.0°C (752.0 mmHg)
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 500 g
Beilstein 21, IV, 1135
Merck Index 15, 4881
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone,chloroform and,benzene
Formula Weight 145.16
Grade ACS Reagent
Insoluble Matter 0.05% max. (in alcohol)
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 8-Hydroxyquinoline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 8-Hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.