missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, 99+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10570531 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10570531 100 g Glass Jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10570531 Supplier Fisher Chemical Supplier No. H/2500/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 148-24-3
Melting Point 72°C
Boiling Point 267°C
Packaging Glass Jar
Molecular Formula C9H7NO
MDL Number MFCD00006807
Quantity 100 g
Synonym 8-hydroxyquinoline, 8-quinolinol, oxyquinoline, oxine, quinophenol, 8-quinol, 8-oxyquinoline, phenopyridine, oxychinolin, bioquin
InChI Key MCJGNVYPOGVAJF-UHFFFAOYSA-N
SMILES OC1=C2N=CC=CC2=CC=C1
IUPAC Name quinolin-8-ol
Molecular Weight (g/mol) 145.16
PubChem CID 1923
ChEBI CHEBI:48981
Formula Weight 145.16g/mol
Physical Form Solid
Show More Show Less
compliance-icons
Product Identifier
  • 8-Hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.