missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinaldine (8-Hydroxy-2-methylquinoline), Mikromol™

Product Code. 30463779
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463779 4 x 25 mg 100mg
30049537 25 mg 25mg
30050289 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463779 Supplier Mikromol Supplier No. MM0390.0100254AMP
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

8-Hydroxyquinaldine (8-Hydroxy-2-methylquinoline), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Hydroxyquinaldine (8-Hydroxy-2-methylquinoline)
CAS 826-81-3
Analyte or Component Names 8-Hydroxyquinaldine (8-Hydroxy-2-methylquinoline)
Shipping Condition Room Temperature
Molecular Formula C10 H9 N O
Certifications/Compliance ISO 17025
Synonym 8-Hydroxyquinaldine, 8-Hydroxy-2-methylquinoline
InChI Formula InChI=1S/C10H9NO/c1-7-5-6-8-3-2-4-9(12)10(8)11-7/h2-6,12H,1H3
Solution Type Neat
SMILES Cc1ccc2cccc(O)c2n1
IUPAC Name 2-methylquinolin-8-ol
Molecular Weight (g/mol) 159.18
Quantity 4 x 25 mg
Formula Weight 159.0684 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.