missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxypyrene-1,3,6-trisulfonic Acid Trisodium Salt (Technical Grade), TRC

Product Code. 30402113
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30402113 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402113 Supplier Toronto Research Chemicals Supplier No. TRCH9537655G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Solvent Green 7
CAS 6358-69-6
Molecular Formula C16 H7 O10 S3 . 3 Na
Quantity 5 g
Synonym 1,3,6-Pyrenetrisulfonic acid, 8-hydroxy-, sodium salt (1:3), 1,3,6-Pyrenetrisulfonic acid, 8-hydroxy-, trisodium salt, 11389 Green, 8-Hydroxy-1,3,6-pyrenetrisulfonic acid trisodium salt, C.I. 59040, C.I. Solvent Green 7, D and C Green No. 8, D&C Green 8, D&C Green No. 8, Fluka 56360, Green 204, Green No. 204, HPTS, Japan Green 204, Japan Green No. 204, Keyacid Pyranine 10G, Pyranine, Pyranine 10G, Pyranine 120, Pyranine Concentrated, Pyranine conk, Pyrene 1, Sanolin Pyranine Green, Solvent Green 7, Trisodium 1-hydroxy-3,6,8-pyrenetrisulfonate, Trisodium 8-hydroxypyrene-1,3,6-trisulfonate, Yellow Pyracide G
InChI Formula InChI=1S/C16H10O10S3.3Na/c17-11-5-12(27(18,19)20)8-3-4-10-14(29(24,25)26)6-13(28(21,22)23)9-2-1-7(11)15(8)16(9)10;;;/h1-6,17H,(H,18,19,20)(H,21,22,23)(H,24,25,26);;;/q;3*+1/p-3
SMILES [Na+].[Na+].[Na+].Oc1cc(c2ccc3c(cc(c4ccc1c2c34)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name trisodium;8-hydroxypyrene-1,3,6-trisulfonate
Molecular Weight (g/mol) 524.39
Formula Weight 523.8894
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.