missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy Moxifloxacin Hydrochloride Salt, TRC

Product Code. 30392167
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392167 50 mg 50mg
30400224 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392167 Supplier Toronto Research Chemicals Supplier No. TRCH94848850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2252446-71-0
Molecular Formula C20 H22 F N3 O4 . Cl H
Molecular Weight (g/mol) 423.8657
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-8-hydroxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-, hydrochloride (1:1), 1-Cyclopropyl-6-fluoro-8-hydroxy-1,4-dihydro-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid hydrochloride, 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-hydroxy-4-oxoquinoline-3-carboxylic acid hydrochloride
IUPAC Name 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-hydroxy-4-oxoquinoline-3-carboxylic acid;hydrochloride
SMILES Cl.OC(=O)C1=CN(C2CC2)c3c(O)c(N4C[C@@H]5CCCN[C@@H]5C4)c(F)cc3C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H22FN3O4.ClH/c21-14-6-12-16(24(11-3-4-11)8-13(18(12)25)20(27)28)19(26)17(14)23-7-10-2-1-5-22-15(10)9-23;/h6,8,10-11,15,22,26H,1-5,7,9H2,(H,27,28);1H/t10-,15+;/m0./s1
Formula Weight 423.1361
Chemical Name or Material Moxifloxacin hydrochloride impurity E
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.