missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy Moxifloxacin Hydrobromide, TRC

Product Code. 30390380
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390380 50 mg
30406118 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390380 Supplier Toronto Research Chemicals Supplier No. TRCH94848550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1292904-74-5
Molecular Formula C20H22FN3O4 . (HBr)
Molecular Weight (g/mol) 468.3
Synonym 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-hydroxy-7-[(4aS,7aS)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic Acid Hydrobromide, Moxifloxacin Imp. E (EP/BP) as HBr salt
IUPAC Name 7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-hydroxy-4-oxoquinoline-3-carboxylic acid;hydrobromide
SMILES Br.OC(=O)C1=CN(C2CC2)c3c(O)c(N4C[C@@H]5CCCN[C@@H]5C4)c(F)cc3C1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H22FN3O4.BrH/c21-14-6-12-16(24(11-3-4-11)8-13(18(12)25)20(27)28)19(26)17(14)23-7-10-2-1-5-22-15(10)9-23;/h6,8,10-11,15,22,26H,1-5,7,9H2,(H,27,28);1H/t10-,15+;/m0./s1
Formula Weight 467.0856
Chemical Name or Material 8-Hydroxy Moxifloxacin Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.