missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Hydroxy Guanosine 5'-Triphosphate Triethylammonium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Hydroxy Guanosine 5'-Triphosphate Triethylammonium Salt |
| Molecular Formula | C10 H16 N5 O15 P3 . (C6 H15 N)n |
| Quantity | 1 mg |
| Synonym | 7,8-Dihydro-8-oxoguanosine 5'-(Tetrahydrogen Triphosphate) Triethylammonium Salt, 2-Amino-9-β-D-ribofuranosylpurine-6,8(1H,9H)-dione 5'-(Tetrahydrogen Triphosphate) Triethylammonium Salt, 8-Ketoguanosine 5'-Triphosphate Bis(triethylammonium) Triethylammonium Salt, 8-OH-GTP Di-TEA Triethylammonium Salt |
| InChI Formula | InChI=1S/C10H16N5O15P3.C6H15N/c11-9-13-6-3(7(18)14-9)12-10(19)15(6)8-5(17)4(16)2(28-8)1-27-32(23,24)30-33(25,26)29-31(20,21)22;1-4-7(5-2)6-3/h2,4-5,8,16-17H,1H2,(H,12,19)(H,23,24)(H,25,26)(H2,20,21,22)(H3,11,13,14,18);4-6H2,1-3H3/t2-,4-,5-,8-;/m1./s1 |
| SMILES | CCN(CC)CC.NC1=Nc2c(nc(O)n2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[C@@H](O)[C@H]3O)C(=O)N1 |
| IUPAC Name | [[(2R,3S,4R,5R)-5-(2-amino-8-hydroxy-6-oxo-1H-purin-9-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-hydroxy-phosphoryl] phosphono hydrogen phosphate;N,N-diethylethanamine |
| Molecular Weight (g/mol) | 640.4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?