missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-7-methoxyflavone, TRC

Product Code. 30404052
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404052 50 mg -
30393476 100 mg 100mg
30393071 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30404052 Supplier Toronto Research Chemicals Supplier No. TRCH58231050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40316-76-5
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.27
Synonym 8-Hydroxy-7-methoxy-2-phenyl-4H-1-benzopyran-4-one, 8-Hydroxy-7-methoxy-2-phenyl-4H-chromen-4-one, 8-Hydroxy-7-methoxyflavone
IUPAC Name 8-hydroxy-7-methoxy-2-phenylchromen-4-one
SMILES COc1ccc2C(=O)C=C(Oc2c1O)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H12O4/c1-19-13-8-7-11-12(17)9-14(20-16(11)15(13)18)10-5-3-2-4-6-10/h2-9,18H,1H3
Formula Weight 268.0736
Chemical Name or Material 8-Hydroxy-7-methoxyflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.