missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-7-iodoquinoline-5-sulfonic Acid, TRC

Product Code. 30400943
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400943 250 mg -
30406461 1 g 1g
30406903 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400943 Supplier Toronto Research Chemicals Supplier No. TRCH943678250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-91-1
Molecular Formula C9H6NO4SI
Molecular Weight (g/mol) 351.11
Synonym 8-Hydroxy-7-iodo-5-quinolinesulfonic Acid, 5-Sulfo-7-iodo-8-hydroxyquinoline, 5-Sulfo-7-iodo-8-quinolinol, 7-Iodo-5-sulfonic acid-8-hydroxyquinoline, 7-Iodo-8-hydroxylquinoline-5-sulfonic Acid, 7-Iodo-8-hydroxyquinoline-5-sulfonic Acid, 7-Iodo-8-quinolinol-5-sulfonic Acid, 7-Iodooxine-5-sulfonic Acid, 8-Hydroxy-7-iodoquinoline-5-sulfonic Acid, Ferron, Ferron (analytical reagent), Loretin, Meditrene, NSC 3784, Quiniophen, Yellon
IUPAC Name 8-hydroxy-7-iodoquinoline-5-sulfonic acid
SMILES Oc1c(I)cc(c2cccnc12)S(=O)(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H6INO4S/c10-6-4-7(16(13,14)15)5-2-1-3-11-8(5)9(6)12/h1-4,12H,(H,13,14,15)
Formula Weight 350.9062
Chemical Name or Material 8-Hydroxy-7-iodoquinoline-5-sulfonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.