missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-5-methyloctanoic Acid, TRC

Product Code. 30399036
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399036 2.5 g 2.5g
30406241 1 g 1g
30405165 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399036 Supplier Toronto Research Chemicals Supplier No. TRCH9502702.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Hydroxy-5-methyloctanoic Acid
Analyte or Component Names 8-Hydroxy-5-methyloctanoic Acid
Shipping Condition Room Temperature
Molecular Formula C9 H18 O3
InChI Formula InChI=1S/C9H18O3/c1-8(5-3-7-10)4-2-6-9(11)12/h8,10H,2-7H2,1H3,(H,11,12)
Solution Type Neat
SMILES CC(CCCO)CCCC(=O)O
IUPAC Name 8-hydroxy-5-methyloctanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 174.237
Quantity 2.5 g
Formula Weight 174.126
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.