missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Hydroxy-3-(hydroxymethyl)-1-anthraquinonyl Glucopyranoside Pentaacetate, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Hydroxy-3-(hydroxymethyl)-1-anthraquinonyl Glucopyranoside Pentaacetate |
| Analyte or Component Names | 8-Hydroxy-3-(hydroxymethyl)-1-anthraquinonyl Glucopyranoside Pentaacetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H20 Cl N O2 . HBr |
| InChI Formula | InChI=1S/C19H20ClNO2.BrH/c1-2-9-21-10-8-14-15(11-17(22)19(23)18(14)20)16(12-21)13-6-4-3-5-7-13;/h2-7,11,16,22-23H,1,8-10,12H2;1H |
| Solution Type | Neat |
| SMILES | Br.Oc1cc2C(CN(CC=C)CCc2c(Cl)c1O)c3ccccc3 |
| IUPAC Name | 6-chloro-1-phenyl-3-prop-2-enyl-1,2,4,5-tetrahydro-3-benzazepine-7,8-diol;hydrobromide |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 410.733 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?