missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-3-(hydroxymethyl)-1-anthraquinonyl Glucopyranoside Pentaacetate, TRC

Product Code. 30396211
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396211 100 mg 100mg
30399843 50 mg 50mg
30399625 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396211 Supplier Toronto Research Chemicals Supplier No. TRCH939810100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Hydroxy-3-(hydroxymethyl)-1-anthraquinonyl Glucopyranoside Pentaacetate
Analyte or Component Names 8-Hydroxy-3-(hydroxymethyl)-1-anthraquinonyl Glucopyranoside Pentaacetate
Shipping Condition Room Temperature
Molecular Formula C19 H20 Cl N O2 . HBr
InChI Formula InChI=1S/C19H20ClNO2.BrH/c1-2-9-21-10-8-14-15(11-17(22)19(23)18(14)20)16(12-21)13-6-4-3-5-7-13;/h2-7,11,16,22-23H,1,8-10,12H2;1H
Solution Type Neat
SMILES Br.Oc1cc2C(CN(CC=C)CCc2c(Cl)c1O)c3ccccc3
IUPAC Name 6-chloro-1-phenyl-3-prop-2-enyl-1,2,4,5-tetrahydro-3-benzazepine-7,8-diol;hydrobromide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 410.733
Quantity 100 mg
Formula Weight 409.044
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.